Přejít k obsahu
Merck
  • Allogeneic ADSCs Induce the Production of Alloreactive Memory-CD8 T Cells through HLA-ABC Antigens.
Allogeneic ADSCs Induce the Production of Alloreactive Memory-CD8 T Cells through HLA-ABC Antigens.
Cells (2020-05-24)
Sung-Ho Chang, Hyun Je Kim, Chung-Gyu Park
ANOTACE
We investigated the immunogenicity of allogeneic human adipose-derived mesenchymal stem cells (ADSCs) through the production of alloreactive-CD8 T and -memory CD8 T cells, based on their human leukocyte antigen (HLA) expression. In surface antigen analysis, ADSCs do not express co-stimulatory molecules, but expresses HLA-ABC, which is further increased by exposure to the pro-inflammatory cytokines as well as IFN-γ alone. For immunogenicity analysis, allogeneic ADSCs cultured in xenofree medium (XF-ADSCs) were incubated with the recipient immune cells for allogeneic-antigen stimulation. As a result, XF-ADSCs induced IFN-γ and IL-17A release by alloreactive-CD8 T cells and the production of alloreactive-CD8 T cell through a direct pathway, although they have immunomodulatory activity. In the analysis of alloreactive memory CD8 T cells, XF-ADSCs also significantly induced the production of CFSE-low-CD8 TEM and -CD8 TCM cells. However, HLA-blocking antibodies significantly inhibited the production of CFSE-low memory-CD8 T cells, indicating that HLAs are the main antigens responsible for the development of allogeneic ADSCs' immunogenicity. These results suggested that HLA surface antigens expressed in allogeneic MSCs should be solved in order to address concerns related to the immunogenicity problem.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Millipore
Multiscreen® 96 well Plate, hydrophobic PVDF membrane, pore size 0.45 μm, sterile