Přejít k obsahu
Merck
  • Toxicokinetics of bisphenol-S and its glucuronide in plasma and urine following oral and dermal exposure in volunteers for the interpretation of biomonitoring data.
Toxicokinetics of bisphenol-S and its glucuronide in plasma and urine following oral and dermal exposure in volunteers for the interpretation of biomonitoring data.
Environment international (2020-03-18)
Imèn Khmiri, Jonathan Côté, Marc Mantha, Rania Khemiri, Marlène Lacroix, Clémence Gely, Pierre-Louis Toutain, Nicole Picard-Hagen, Véronique Gayrard, Michèle Bouchard
ANOTACE
The measurement of bisphenol-S (BPS) and its glucurono-conjugate (BPSG) in urine may be used for the biomonitoring of exposure in populations. However, this requires a thorough knowledge of their toxicokinetics. The time courses of BPS and BPSG were assessed in accessible biological matrices of orally and dermally exposed volunteers. Under the approval of the Research Ethics Committee of the University of Montreal, six volunteers were orally exposed to a BPS-d8 deuterated dose of 0.1 mg/kg body weight (bw). One month later, 1 mg/kg bw of BPS-d8 were applied on 40 cm2 of the forearm and then washed 6 h after application. Blood samples were taken prior to dosing and at fixed time periods over 48 h after treatment; complete urine voids were collected pre-exposure and at pre-established intervals over 72 h postdosing. Following oral exposure, the plasma concentration-time courses of BPS-d8 and BPSG-d8 over 48 h evolved in parallel, and showed a rapid appearance and elimination. Average peak values (±SD) were reached at 0.7 ± 0.1 and 1.1 ± 0.4 h postdosing and mean (±SD) apparent elimination half-lives (t½) of 7.9 ± 1.1 and 9.3 ± 7.0 h were calculated from the terminal phase of BPS-d8 and BPSG-d8 in plasma, respectively. The fraction of BPS-d8 reaching the systemic circulation unchanged (i.e. bioavailability) was further estimated at 62 ± 5% on average (±SD) and the systemic plasma clearance at 0.57 ± 0.07 L/kg bw/h. Plasma concentration-time courses and urinary excretion rate profiles roughly evolved in parallel for both substances, as expected. The average percent (±SD) of the administered dose recovered in urine as BPS-d8 and BPSG-d8 over the 0-72 h period postdosing was 1.72 ± 1.3 and 54 ± 10%. Following dermal application, plasma levels were under the lower limit of quantification (LLOQ) at most time points. However, peak values were reached between 5 and 8 h depending on individuals, suggesting a slower absorption rate compared to oral exposure. Similarly, limited amounts of BPS-d8 and its conjugate were recovered in urine and peak excretion rates were reached between 5 and 11 h postdosing. The average percent (±SD) of the administered dose recovered in urine as BPS-d8 and BPSG-d8 was about 0.004 ± 0.003 and 0.09 ± 0.07%, respectively. This study provided greater precision on the kinetics of this contaminant in humans and, in particular, evidenced major differences between BPA and BPS kinetics with much higher systemic levels of active BPS than BPA, an observation explained by a higher oral bioavailability of BPS than BPA. These data should also be useful in developing a toxicokinetic model for a better interpretation of biomonitoring data.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%