Přejít k obsahu
Merck
  • Heterocyclic aromatic amine concentrations and quality characteristics of traditional smoked and roasted poultry products on the northern Chinese market.
Heterocyclic aromatic amine concentrations and quality characteristics of traditional smoked and roasted poultry products on the northern Chinese market.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2019-11-05)
Lang Zhang, Hongzhen Du, Pin Zhang, Baohua Kong, Qian Liu
ANOTACE
This study evaluated the heterocyclic aromatic amine (HAA) contents and quality characteristics of seven kinds of traditional smoked and roasted poultry products on the northern Chinese market. Harbin smoked chicken had the most abundant total HAAs, followed by Haroulian roasted chicken and Yishou smoked chicken. The contents of Norharman and Harman were much higher than those of other kinds of HAAs (P < 0.05). The water content of samples varied from 59.01% to 69.98% and the water activity varied from 0.953 to 0.976. The carbonyl content and TBARS values of the Beijing roasted duck and the Duiqing roasted goose were much higher than those of the other samples (P < 0.05). The sensory evaluation result of the Beijing roasted chicken was higher than that of the other samples (P < 0.05). Overall, the levels of HAAs in the industrial smoked and roasted products were lower than those in non-industrial products, which may provide a theoretical basis for the industrial production of smoked and roasted poultry products.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
2-Amino-5-phenylpyridine, AldrichCPR