Přejít k obsahu
Merck
  • Estrogen's Effects on Excitatory Synaptic Transmission Entail Integrin and TrkB Transactivation and Depend Upon β1-integrin function.
Estrogen's Effects on Excitatory Synaptic Transmission Entail Integrin and TrkB Transactivation and Depend Upon β1-integrin function.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2016-06-09)
Weisheng Wang, Svetlana Kantorovich, Alex H Babayan, Bowen Hou, Christine M Gall, Gary Lynch
ANOTACE
Estradiol (E2) perfusion rapidly increases the strength of fast excitatory transmission and facilitates long-term potentiation in the hippocampus, two effects likely related to its memory-enhancing properties. Past studies showed that E2's facilitation of transmission involves activation of RhoA signaling leading to actin polymerization in dendritic spines. Here we report that brief exposure of adult male hippocampal slices to 1 nM E2 increases the percentage of postsynaptic densities associated with high levels of immunoreactivity for activated forms of the BDNF receptor TrkB and β1-integrins, two synaptic receptors that engage actin regulatory RhoA signaling. The effects of E2 on baseline synaptic responses were unaffected by pretreatment with the TrkB-Fc scavenger for extracellular BDNF or TrkB antagonism, but were eliminated by neutralizing antisera for β1-integrins. E2 effects on synaptic responses were also absent in conditional β1-integrin knockouts, and with inhibition of matrix metalloproteinases, extracellular enzymes that generate integrin ligands. We propose that E2, acting through estrogen receptor-β, transactivates synaptic TrkB and β1-integrin, and via mechanisms dependent on integrin activation and signaling, reversibly reorganizes the spine cytoskeleton and thereby enhances synaptic responses in adult hippocampus.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4C10, azide free, clone P4C10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, activated, clone B44, azide free, clone B44, Chemicon®, from mouse