- An induced pluripotent stem cell line (TRNDi010-C) from a patient carrying a homozygous p.R401X mutation in the NGLY1 gene.
An induced pluripotent stem cell line (TRNDi010-C) from a patient carrying a homozygous p.R401X mutation in the NGLY1 gene.
Stem cell research (2019-07-22)
Shu Yang, Yu-Shan Cheng, Rong Li, Manisha Pradhan, Junjie Hong, Jeanette Beers, Jizhong Zou, Chengyu Liu, Matt Might, Steven Rodems, Wei Zheng
PMID31326749
ANOTACE
NGLY1 deficiency is a rare inherited disorder caused by mutations in the NGLY1 gene encoding N-glycanase 1 that is a hydrolase for N-linked glycosylated proteins. An induced pluripotent stem cell (iPSC) line was generated from the dermal fibroblasts of a 16-year-old patient with homozygous mutation of p.R401X (c.1201 A>T) in the NGLY1 gene. Our iPSC model offers a useful resource to study the disease pathophysiology and to develop therapeutics for treatment of NGLY1 patients.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit