Přejít k obsahu
Merck
The HER3 pathway as a potential target for inhibition in patients with biliary tract cancers.
PloS one (2018-10-20)
Angela Lamarca, Salvatore Galdy, Jorge Barriuso, Sharzad Moghadam, Elizabeth Beckett, Jane Rogan, Alison Backen, Catherine Billington, Mairéad G McNamara, Richard A Hubner, Angela Cramer, Juan W Valle
ANOTACE
Expression of human epidermal growth factor receptor (HER)2 and HER3 have been investigated in small BTC studies using variable scoring systems. HER2 and HER3 overexpression/amplification were explored following internationally agreed guidelines using immunohistochemistry (IHC) and fluorescent in-situ hybridisation (FISH), respectively. Logistic regression and survival analysis (Kaplan Meier, Log rank test and Cox Regression) were used for statistical analysis. Sixty-seven eligible patients with Stage I/II (31.3%) or III/IV (68.7%) disease at diagnosis were included. Membrane HER2 overexpression/amplification was identified in 1 patient (1%). HER3 overexpression was predominantly cytoplasmic; the rate of overexpression/amplification of HER3 in membrane and cytoplasm was 16% [ampullary cancer (AMP) (1/13; 8%), gallbladder cancer (GBC) (1/10; 10%), intra-hepatic cholangiocarcinoma (ICC) (6/26; 23%), extra-hepatic cholangiocarcinoma (ECC) (3/18; 17%)] and 24% [AMP (1/13; 8%), GBC (1/10; 10%), ICC (10/26; 38%), ECC (4/18; 22%)], respectively. A significant subset of patients with BTC expressed HER3. Inhibition of HER3 warrants further investigation. A better understanding of the downstream effects of HER3 in BTC requires further mechanistic investigations to identify new biomarkers and improve patient selection for future clinical trials.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-erbB-3/HER-3 Antibody, clone 2F12, clone 2F12, Upstate®, from mouse