Přejít k obsahu
Merck
  • Palladium-catalyzed cross-coupling of electron-poor terminal alkynes with arylboronic acids under ligand-free and aerobic conditions.
Palladium-catalyzed cross-coupling of electron-poor terminal alkynes with arylboronic acids under ligand-free and aerobic conditions.
The Journal of organic chemistry (2010-08-14)
Ming-Bo Zhou, Wen-Ting Wei, Ye-Xiang Xie, Yong Lei, Jin-Heng Li
ANOTACE
Palladium-catalyzed cross-coupling reaction of terminal alkynes with arylboronic acids has been described. In the presence of Pd(OAc)(2) and Ag(2)O, a variety of terminal alkynes, including electron-poor terminal alkynes, smoothly underwent the reaction with numerous boronic acids to afford the corresponding internal alkynes in moderate to good yields. Moreover, this methodology was applied to the synthesis of 1H-isochromenes and diynes. It is noteworthy that the reaction proceeds under ligand-free and relative lower loading Pd conditions, and the maximal TONs (turnover numbers) of the reaction are up to 720,000.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
trans-1-Propen-1-ylboronic acid, ≥95.0%