Přejít k obsahu
Merck
Reductive Cyclopropanations Catalyzed by Dinuclear Nickel Complexes.
Angewandte Chemie (International ed. in English) (2016-01-30)
You-Yun Zhou, Christopher Uyeda
ANOTACE
Dinuclear Ni complexes supported by naphthyridine-diimine (NDI) ligands catalyze the reductive cyclopropanation of alkenes with CH2 Cl2 as the methylene source. The use of mild terminal reductants (Zn or Et2 Zn) confers significant functional-group tolerance, and the catalyst accommodates structurally and electronically diverse alkenes. Mononickel catalysts bearing related N chelates afford comparatively low cyclopropane yields (≤20 %). These results constitute an entry into catalytic carbene transformations from oxidized methylene precursors.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
iPr-NDI, ≥95%