Přejít k obsahu
Merck
Catalytic Azoarene Synthesis from Aryl Azides Enabled by a Dinuclear Ni Complex.
Journal of the American Chemical Society (2018-03-01)
Ian G Powers, John M Andjaba, Xuyi Luo, Jianguo Mei, Christopher Uyeda
ANOTACE
Azoarenes are valuable chromophores that have been extensively incorporated as photoswitchable elements in molecular machines and biologically active compounds. Here, we report a catalytic nitrene dimerization reaction that provides access to structurally and electronically diverse azoarenes. The reaction utilizes aryl azides as nitrene precursors and generates only gaseous N2 as a byproduct. By circumventing the use of a stoichiometric redox reagent, a broad range of organic functional groups are tolerated, and common byproducts of current methods are avoided. A catalyst featuring a Ni-Ni bond is found to be uniquely effective relative to those containing only a single Ni center. The mechanistic origins of this nuclearity effect are described.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
iPr-NDI, ≥95%