Přejít k obsahu
Merck
  • SIRT7 mediates L1 elements transcriptional repression and their association with the nuclear lamina.
SIRT7 mediates L1 elements transcriptional repression and their association with the nuclear lamina.
Nucleic acids research (2019-06-22)
Berta N Vazquez, Joshua K Thackray, Nicolas G Simonet, Sanjay Chahar, Noriko Kane-Goldsmith, Simon J Newkirk, Suman Lee, Jinchuan Xing, Michael P Verzi, Wenfeng An, Alejandro Vaquero, Jay A Tischfield, Lourdes Serrano
ANOTACE
Long interspersed elements-1 (LINE-1, L1) are retrotransposons that hold the capacity of self-propagation in the genome with potential mutagenic outcomes. How somatic cells restrict L1 activity and how this process becomes dysfunctional during aging and in cancer cells is poorly understood. L1s are enriched at lamin-associated domains, heterochromatic regions of the nuclear periphery. Whether this association is necessary for their repression has been elusive. Here we show that the sirtuin family member SIRT7 participates in the epigenetic transcriptional repression of L1 genome-wide in both mouse and human cells. SIRT7 depletion leads to increased L1 expression and retrotransposition. Mechanistically, we identify a novel interplay between SIRT7 and Lamin A/C in L1 repression. Our results demonstrate that SIRT7-mediated H3K18 deacetylation regulates L1 expression and promotes L1 association with elements of the nuclear lamina. The failure of such activity might contribute to the observed genome instability and compromised viability in SIRT7 knockout mice. Overall, our results reveal a novel function of SIRT7 on chromatin organization by mediating the anchoring of L1 to the nuclear envelope, and a new functional link of the nuclear lamina with transcriptional repression.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human LMNA