Přejít k obsahu
Merck
Identification of a cellularly active SIRT6 allosteric activator.
Nature chemical biology (2018-10-31)
Zhimin Huang, Junxing Zhao, Wei Deng, Yingyi Chen, Jialin Shang, Kun Song, Lu Zhang, Chengxiang Wang, Shaoyong Lu, Xiuyan Yang, Bin He, Jinrong Min, Hao Hu, Minjia Tan, Jianrong Xu, Qiufen Zhang, Jie Zhong, Xiaoxiang Sun, Zhiyong Mao, Houwen Lin, Mingzhe Xiao, Y Eugene Chin, Hualiang Jiang, Ying Xu, Guoqiang Chen, Jian Zhang
ANOTACE
SIRT6, a member of the SIRT deacetylase family, is responsible for deacetylation of histone H3 Nε-acetyl-lysines 9 (H3K9ac) and 56 (H3K56ac). As a tumor suppressor, SIRT6 has frequently been found to have low expression in various cancers. Here, we report the identification of MDL-800, a selective SIRT6 activator. MDL-800 increased the deacetylase activity of SIRT6 by up to 22-fold via binding to an allosteric site; this interaction led to a global decrease in H3K9ac and H3K56ac levels in human hepatocellular carcinoma (HCC) cells. Consequently, MDL-800 inhibited the proliferation of HCC cells via SIRT6-driven cell-cycle arrest and was effective in a tumor xenograft model. Together, these data demonstrate that pharmacological activation of SIRT6 is a potential therapeutic approach for the treatment of HCC. MDL-800 is a first-in-class small-molecule cellular SIRT6 activator that can be used to physiologically and pathologically investigate the roles of SIRT6 deacetylation.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
MDL-800, ≥98% (HPLC)