Přejít k obsahu
Merck
  • Specificity of inhibitors of serine palmitoyltransferase (SPT), a key enzyme in sphingolipid biosynthesis, in intact cells. A novel evaluation system using an SPT-defective mammalian cell mutant.
Specificity of inhibitors of serine palmitoyltransferase (SPT), a key enzyme in sphingolipid biosynthesis, in intact cells. A novel evaluation system using an SPT-defective mammalian cell mutant.
Biochemical pharmacology (2000-03-29)
K Hanada, M Nishijima, T Fujita, S Kobayashi
ANOTACE
In the present study, we demonstrate a model cell system for evaluating the specificity of inhibitors of serine palmitoyltransferase (SPT), the enzyme that catalyzes the first step of sphingolipid biosynthesis. The LY-B strain is a Chinese hamster ovary (CHO) cell mutant defective in SPT, and the LY-B/cLCB1 strain is a genetically corrected revertant of the mutant. Although LY-B cells grew only slightly in sphingolipid-deficient medium, their growth was restored to the level of LY-B/cLCB1 cells under sphingosine-supplied conditions, indicating that, in CHO cells, the growth inhibition caused by SPT inactivation was rescued almost fully by the metabolic complementation of sphingolipids. Cultivation of LY-B/cLCB1 cells in sphingolipid-deficient medium in the presence of 10 microM sphingofungin B and ISP-1 (myriocin, thermozymocidin), potent inhibitors of SPT activity, caused severe growth inhibition with approximately 95% inhibition of de novo sphingolipid synthesis. The growth inhibition by sphingofungin B and ISP-1 was rescued substantially by exogenous sphingosine, whereas the cytotoxicity of two other types of SPT inhibitor, L-cycloserine and beta-chloro-L-alanine, was hardly rescued. Similar cytotoxic patterns of these inhibitors also were observed on the growth of SPT-defective LY-B cells cultured under sphingosine-supplied conditions. The SPT inhibitors did not affect metabolic conversion of exogenous [(3)H]sphingosine to complex sphingolipids. Thus, the cytotoxicity of sphingofungin B and ISP-1, but not L-cycloserine or beta-chloro-L-alanine, is due largely to inhibition of sphingolipid synthesis by inhibiting the SPT activity.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Myriocin, Mycelia sterilia, A potent immunosuppressant.