Přejít k obsahu
Merck
Near Room Temperature Light-Activated WS2-Decorated rGO as NO2 Gas Sensor.
Sensors (Basel, Switzerland) (2019-06-12)
Valentina Paolucci, Seyed Mahmoud Emamjomeh, Luca Ottaviano, Carlo Cantalini
ANOTACE
The NO2 response in the range of 200 ppb to 1 ppm of a chemoresistive WS2-decorated rGO sensor has been investigated at operating temperatures of 25 °C and 50 °C in dry and humid air (40% RH) under dark and Purple Blue (PB) light conditions (λ = 430 nm). Few-layers WS2, exfoliated by ball milling and sonication technique, with average dimensions of 200 nm, have been mixed with rGO flakes (average dimension 700 nm) to yield WS2-decorated rGO, deposited on Si3N4 substrates, provided with platinum (30 μm gap distance) finger-type electrodes. TEM analysis showed the formation of homogeneous and well-dispersed WS2 flakes distributed over a thin, continuous and uniform underlying layer of interconnected rGO flakes. XPS and STEM revealed a partial oxidation of WS2 flakes leading to the formation of 18% amorphous WO3 over the WS2 flakes. PB-light irradiation and mild heating of the sensor at 50 °C substantially enhanced the baseline recovery yielding improved adsorption/desorption rates, with detection limit of 400 ppb NO2 and reproducible gas responses. Cross sensitivity tests with humid air interfering vapor highlighted a negligible influence of water vapor on the NO2 response. A charge carrier mechanism between WS2 and rGO is proposed and discussed to explain the overall NO2 and H2O response of the WS2-rGO hybrids.