Přejít k obsahu
Merck
A safe and non-flammable sodium metal battery based on an ionic liquid electrolyte.
Nature communications (2019-07-26)
Hao Sun, Guanzhou Zhu, Xintong Xu, Meng Liao, Yuan-Yao Li, Michael Angell, Meng Gu, Yuanmin Zhu, Wei Hsuan Hung, Jiachen Li, Yun Kuang, Yongtao Meng, Meng-Chang Lin, Huisheng Peng, Hongjie Dai
ANOTACE
Rechargeable sodium metal batteries with high energy density could be important to a wide range of energy applications in modern society. The pursuit of higher energy density should ideally come with high safety, a goal difficult for electrolytes based on organic solvents. Here we report a chloroaluminate ionic liquid electrolyte comprised of aluminium chloride/1-methyl-3-ethylimidazolium chloride/sodium chloride ionic liquid spiked with two important additives, ethylaluminum dichloride and 1-ethyl-3-methylimidazolium bis(fluorosulfonyl)imide. This leads to the first chloroaluminate based ionic liquid electrolyte for rechargeable sodium metal battery. The obtained batteries reached voltages up to ~ 4 V, high Coulombic efficiency up to 99.9%, and high energy and power density of ~ 420 Wh kg-1 and ~ 1766 W kg-1, respectively. The batteries retained over 90% of the original capacity after 700 cycles, suggesting an effective approach to sodium metal batteries with high energy/high power density, long cycle life and high safety.