Přejít k obsahu
Merck
  • Evaluation of Single and Joint Toxicity of Perfluorinated Carboxylic Acids and Copper to Metal-Resistant Arthrobacter Strains.
Evaluation of Single and Joint Toxicity of Perfluorinated Carboxylic Acids and Copper to Metal-Resistant Arthrobacter Strains.
International journal of environmental research and public health (2019-01-10)
Yanping Cai, Haiyan Chen, Huilun Chen, Haiqing Li, Shuo Yang, Fei Wang
ANOTACE
Perfluorocarboxylic acid compounds (PFCAs) and copper have been regarded as ubiquitous environmental contaminants in aquatic ecosystems worldwide. However, data on their possible joint toxic effects on microorganisms are still lacking. To study the combined effects of four PFCAs with different carbon chain lengths and copper, a series of experiments were conducted to explore the acute toxicity of these PFCAs in the absence and presence of copper on a metal-resistant Arthrobacter strain GQ-9 by microcalorimetry. The thermokinetic parameters, including growth rate constant (k), inhibitory ratio (I), and half inhibitory concentration (IC50), were calculated and compared using the data obtained from the power-time curves. Our work revealed that GQ-9 is more resistant to perfluorooctanoic acid (PFOA) than Escherichia coli. The single and joint toxicity of PFCAs with copper are dose- and carbon chain length-dependent. The longer the carbon chain length of PFCAs, the higher the toxicity. In addition, PFCAs interacted synergistically with copper. This work could provide useful information for the risk assessment of co-exposure to perfluorinated compounds and heavy metals in natural environments.