Přejít k obsahu
Merck
  • Selenium Attenuates Staphylococcus aureus Mastitis in Mice by Inhibiting the Activation of the NALP3 Inflammasome and NF-κB/MAPK Pathway.
Selenium Attenuates Staphylococcus aureus Mastitis in Mice by Inhibiting the Activation of the NALP3 Inflammasome and NF-κB/MAPK Pathway.
Biological trace element research (2018-12-14)
Junkun Ma, Shuo Zhu, Yingfang Guo, Meilin Hao, Yu Chen, Ying Wang, Mei Yang, Jianguo Chen, Mengyao Guo
ANOTACE
Mastitis is one of the most important diseases affecting the dairy industry in the world, and it also poses a great threat to human food safety. In this study, we explored whether selenium can inhibit the activation of the NALP3 inflammasome and NF-κB/MAPK pathway to achieve anti-inflammatory effects. Sixty BALB/c female mice were randomly divided into three groups according to diets of different selenium concentrations (high, normal, and low). After 90 days, mice fed the same selenium concentration were randomly divided into two smaller groups, one of which was inoculated with Staphylococcus aureus and the other injected with saline as a control. Through histopathologic examination staining, western blot, qPCR, and ELISA, the results showed that with increasing selenium concentrations, the expression levels of IL-1β, TNF-α, NALP3, caspase-1, and ASC were decreased in mouse mammary tissue. Therefore, this study revealed that selenium can attenuate S. aureus mastitis by inhibiting the activation of the NALP3 inflammasome and NF-κB/MAPK pathway.