Přejít k obsahu
Merck
  • Molecular cloning and functional characterization of TRIF in large yellow croaker Larimichthys crocea.
Molecular cloning and functional characterization of TRIF in large yellow croaker Larimichthys crocea.
Fish & shellfish immunology (2019-05-16)
Peng Fei Zou, Juan Juan Shen, Ying Li, Qingpi Yan, Zhi Hua Zou, Zi Ping Zhang, Yi Lei Wang
ANOTACE
As an adaptor in Toll-like receptor (TLR) signaling pathway, Toll/interleukin-1 receptor (TIR) domain containing adaptor inducing interferon-β (TRIF) mediates downstream signaling cascades and plays important roles in host innate immune responses. In the present study, a TRIF ortholog named Lc-TRIF was identified in large yellow croaker (Larimichthys crocea). Sequence comparison analysis revealed that Lc-TRIF has a conserved TIR domain but without TRAF6 binding motif. The genome structure of Lc-TRIF is conserved, with two exons and one intron. Syntenic comparison showed that the loci of fish TRIF was different from that in mammals or birds, and TRAM was absent in the genomes of fish, amphibians, and birds, but present in mammals and reptiles. Expression analysis revealed that Lc-TRIF was broadly expressed in examined organs/tissues, with the highest expression level in gill and weakest in brain, and could be up-regulated under poly I:C, LPS, PGN, and Pseudomonas plecoglossicida stimulation. Fluorescence microscopy results showed that Lc-TRIF exhibited a global localization throughout the entire cell including the nucleus in HEK 293T cells. Additionally, luciferase assays demonstrated that Lc-TRIF expression could significantly induce NF-κB, type I IFN, IRF3 as well as IRF7 promoter activation. These results collectively indicated that Lc-TRIF was function in host antiviral and antibacterial responses via NF-κB and IRF3/7 related signaling pathway.