Přejít k obsahu
Merck
  • Mass spectrometry imaging guided molecular networking to expedite discovery and structural analysis of agarwood natural products.
Mass spectrometry imaging guided molecular networking to expedite discovery and structural analysis of agarwood natural products.
Analytica chimica acta (2019-08-15)
Ting-Hao Kuo, Hou-Chun Huang, Cheng-Chih Hsu
ANOTACE
Structural analysis of biomolecules is essential to natural product discovery, especially for precious biomaterials such as agarwood. However, one of the greatest challenges to the characterization of natural products is the profound cost in time and manpower to the structural elucidation of these highly diverse compounds. Here, we demonstrate a multi-modal mass spectrometric strategy, integrating matrix-assisted laser desorption ionization (MALDI) mass spectrometry imaging (MSI) and mass spectral molecular networking, to uncover agarwood natural products of Aquilaria sinensis trees. A simple workflow for preparing wood sections for MALDI-MSI analysis was demonstrated. Notably, tens of natural products in the agarwood region in wood stem section of A. sinensis were spatially revealed by MALDI-MSI. For the first time, such a great number of plant specialized metabolites is obtained by a single wood section MSI. Guided by the spatially resolved features, mass spectral molecular networking was subsequently applied for structural analysis of the agarwood natural products, in which three major classes of 2-(2-phenylethyl)chromones and their analogues were putatively characterized. These results suggest an efficient strategy to the dereplication of plant natural products.