Přejít k obsahu
Merck
  • The influence of a new derivate of kisspeptin-10 - Kissorphin (KSO) on the rewarding effects of morphine in the conditioned place preference (CPP) test in male rats.
The influence of a new derivate of kisspeptin-10 - Kissorphin (KSO) on the rewarding effects of morphine in the conditioned place preference (CPP) test in male rats.
Behavioural brain research (2019-06-22)
E Gibula-Tarlowska, E Kedzierska, K Piechura, J Silberring, J H Kotlinska
ANOTACE
Kissorphin (KSO) is a new peptide derived from kisspeptin-10. Previous study has indicated that this peptide displays neuropeptide FF (NPFF)-like anti-opioid activity. Herein, we examined the influence of KSO (1; 3, and 10 nmol, intravenously [i.v.]), on the rewarding action of morphine (5 mg/kg, intraperitoneally [i.p.]), using the unbiased design of the conditioned place preference (CPP) paradigm in rats. To test the effect of KSO on the acquisition of morphine-induced CPP, KSO and morphine were co-injected during conditioning with no drugs treatment on the test day. To investigate the effect of KSO on the expression of morphine-induced CPP, morphine alone was given during the conditioning phase (1 × 3 days) and KSO was administered 5 min prior to the placement in the CPP apparatus on the test day. To estimate the influence of KSO on the reinstatement of morphine-induced CPP, KSO was given 5 min before a priming dose of morphine (5 mg/kg, i.p.) on the reinstatement test day. The results show that KSO inhibited the acquisition, expression and reinstatement of morphine-induced CPP. The strongest effect of KSO was observed at the dose of 10 nmol (acquisition and reinstatement) or 1 nmol (expression). KSO given alone, neither induced place preference, nor aversion. Furthermore, the morphine-modulating effects of KSO were markedly antagonized by pretreatment with RF9 (10 nmol, i.v.), the NPFF receptors selective antagonist. Thus, KSO inhibited the morphine-induced CPP mainly by involving specific activation of NPFF receptors. Overall, these data further support the anti-opioid character of KSO.