Přejít k obsahu
Merck
  • The effect of fludrocortisone on the uterine receptivity partially mediated by ERK1/2-mTOR pathway.
The effect of fludrocortisone on the uterine receptivity partially mediated by ERK1/2-mTOR pathway.
Journal of cellular physiology (2019-04-11)
Mohammad Bakhtiar Hesam Shariati, Abbas Majdi Seghinsara, Naser Shokrzadeh, Behrooz Niknafs
ANOTACE
Implantation of embryos needs endometrial receptivity. Mineralocorticoids is one of the causes influencing the implantation window. This study targeted to evaluation fludrocortisone different properties on endometrial receptivity. The objective of this study was to assess whether treatment with fludrocortisone could impact the expression of diverse genes and proteins that are involved in uterine receptivity in mice. In this study, 40 female adult BALB/c mice were used. The samples were allocated to four groups of ten. Control group (C) received: vehicle; fludrocortisone group (FCA): received 1.5 mg/kg fludrocortisone; PP242 group (PP242): received 30 mg/kg PP242; fludrocortisone+PP242 group (FCA+PP242): received fludrocortisone and PP242. Mice were killed on window implantation day after mating and confirmed pregnancy. The endometrial epithelium of mouse was collected to assess mRNA expression of leukemia inhibitory factor (LIF), mucin-1 (MUC1), heparin-binding epidermal growth factor (HB-EGF), (Msx.1), miRNA Let-7a, and miRNA 223-3p as well as protein expression of extracellular signal-regulated kinase 1/2 (ERK1/2), mammalian target of rapamycin (mTOR), and eukaryotic translation initiation factor 4E-binding protein 1 (4E-BP1) in the uterine using real-time PCR and western blot, respectively. In comparison with the control group, fludrocortisone administration upregulated the expression of LIF, HB-EGF, Msx.1, miRNA Let-7a, ERK1/2, and mTOR in the epithelial endometrium. The PP242-treated group demonstrated a significant rise in the expression of MUC1, miRNA 223-3p and a remarkable decline in ERK1/2 and p-4E-BP1 levels in comparison with the control group. Combination therapy of (FCA+PP242) resulted in a remarkable rise in LIF, Msx-1, HB-EGF, ERK1/2, and mTOR levels, in comparison with the PP242 group. Furthermore, combination therapy of (FCA+PP242) downregulated the expression of MUC1 in comparison with the PP242-treated group. According to the results, fludrocortisone affected uterine receptivity possibly by means of modulating the expression of genes involved in the uterine receptivity and activation of the ERK1/2-mTOR pathway.