Přejít k obsahu
Merck
Synthesis and Characterization of Silicone Contact Lenses Based on TRIS-DMA-NVP-HEMA Hydrogels.
Polymers (2019-06-05)
Nguyen-Phuong-Dung Tran, Ming-Chien Yang
ANOTACE
In this study, silicone-based hydrogel contact lenses were prepared by the polymerization of 3-(methacryloyloxy)propyltris(trimethylsiloxy)silane (TRIS), N,N-dimethylacrylamide (DMA), 1-vinyl-2-pyrrolidinone (NVP), and 2-hydroxyethylmethacrylate (HEMA). The properties of silicone hydrogel lenses were analyzed based on the methods such as equilibrium water content, oxygen permeability, optical transparency, contact angle, mechanical test, protein adsorption, and cell toxicity. The results showed that the TRIS content in all formulations increased the oxygen permeability and decreased the equilibrium water content, while both DMA and NVP contributed the hydrophilicity of the hydrogels. The maximum value of oxygen permeability was 74.9 barrers, corresponding to an equilibrium water content of 44.5% as well as a contact angle of 82°. Moreover, L929 fibroblasts grew on all these hydrogels, suggesting non-cytotoxicity. In general, the silicone hydrogels in this work exhibited good oxygen permeability, stiffness, and optical transparency as well as anti-protein adsorption. Hence, these silicone hydrogel polymers would be feasible for making contact lens.