Přejít k obsahu
Merck
  • Do arachidonic acid metabolites affect apoptosis in bovine endometrial cells with silenced PPAR genes?
Do arachidonic acid metabolites affect apoptosis in bovine endometrial cells with silenced PPAR genes?
Prostaglandins & other lipid mediators (2019-05-22)
A A Szczepańska, M Łupicka, A J Korzekwa
ANOTACE
Peroxisome proliferator-activated receptors (PPARs) are expressed in bovine uterus, and their agonists are arachidonic acid (AA) metabolites. We hypothesised that silencing of PPAR genes in bovine endometrial stromal cells (ESC) would change the intracellular signalling through PPAR and affect apoptosis after cell treatment with different AA metabolites. The study's aims are detection of apoptosis and examining the influence of prostaglandins and leukotrienes on apoptosis occurring in physiological ESC and cells with silenced PPAR (α, δ, and γ) genes. Silencing the PPARα and PPARδ genes in cells resulted in increased DNA fragmentation and mRNA and protein expression of caspase (CASP) -3 and -8 (P < 0.05). Neither DNA fragmentation nor the mRNA and protein expression of CASP3 and -8 in cells with silenced PPARγ gene were changed compared to physiological cells (P > 0.05). Among PPARs, PPARα and PPARδ appear to inhibit apoptosis, and AA metabolites, as PPAR agonists, modify this process in bovine ESC.