Přejít k obsahu
Merck
  • Butyl Methacrylate-Co-Ethylene Glycol Dimethacrylate Monolith for Online in-Tube SPME-UHPLC-MS/MS to Determine Chlopromazine, Clozapine, Quetiapine, Olanzapine, and Their Metabolites in Plasma Samples.
Butyl Methacrylate-Co-Ethylene Glycol Dimethacrylate Monolith for Online in-Tube SPME-UHPLC-MS/MS to Determine Chlopromazine, Clozapine, Quetiapine, Olanzapine, and Their Metabolites in Plasma Samples.
Molecules (Basel, Switzerland) (2019-01-19)
Luiz G M Beloti, Luis F C Miranda, Maria Eugênia C Queiroz
ANOTACE
This manuscript describes a sensitive, selective, and online in-tube solid-phase microextraction coupled with an ultrahigh performance liquid chromatography-tandem mass spectrometry (in-tube SPME-UHPLC-MS/MS) method to determine chlopromazine, clozapine, quetiapine, olanzapine, and their metabolites in plasma samples from schizophrenic patients. Organic poly(butyl methacrylate-co-ethylene glycol dimethacrylate) monolith was synthesized on the internal surface of a fused silica capillary (covalent bonds) for in-tube SPME. Analyte extraction and analysis was conducted by connecting the monolithic capillary to an UHPLC-MS/MS system. The monolith was characterized by scanning electron microscopy (SEM) and Fourier transform infrared spectrometry (FTIR). The developed method presented adequate linearity for all the target antipsychotics: R² was higher than 0.9975, lack-of-fit ranged from 0.115 to 0.955, precision had variation coefficients lower than 14.2%, and accuracy had relative standard error values ranging from -13.5% to 14.6%, with the exception of the lower limit of quantification (LLOQ). The LLOQ values in plasma samples were 10 ng mL-1 for all analytes. The developed method was successfully applied to determine antipsychotics and their metabolites in plasma samples from schizophrenic patients.