Přejít k obsahu
Merck
Identification and characterization of glycosylation sites on Litopenaeus vannamei hemocyanin.
FEBS letters (2019-03-23)
Zehui Zhang, Ruiwei Li, Jude Juventus Aweya, Fan Wang, Mingqi Zhong, Yueling Zhang
ANOTACE
The respiratory glycoprotein hemocyanin has been implicated in immune-related functions. Using lectin blotting, we show that the binding of shrimp (Litopenaeus vannamei) hemocyanin to concanavalin A decreases markedly with O-glycosidase treatment but not with PNGase F. Twelve O-glycosylation sites, three on the large hemocyanin subunit and nine on the small hemocyanin subunit (HMCs), were identified by LC-MS/MS. Importantly, when the glycosylation sites at Thr-537, Ser-539, and Thr-542 on the C terminus of HMCs were replaced with alanine, the resultant mutant hemocyanin had reduced carbohydrate content, coupled with a fourfold reduction in bacterial agglutination and 0.2-fold reduction in antibacterial activities toward Vibrio parahaemolyticus and Staphylococcus aureus. These results suggest that the glycosylation sites on shrimp hemocyanin are closely related to its immunological functions.