Přejít k obsahu
Merck
Mapping Interactions of Microbial Metabolites with Human G-Protein-Coupled Receptors.
Cell host & microbe (2019-08-06)
Dominic A Colosimo, Jeffrey A Kohn, Peter M Luo, Frank J Piscotta, Sun M Han, Amanda J Pickard, Arka Rao, Justin R Cross, Louis J Cohen, Sean F Brady
ANOTACE
Despite evidence linking the human microbiome to health and disease, how the microbiota affects human physiology remains largely unknown. Microbiota-encoded metabolites are expected to play an integral role in human health. Therefore, assigning function to these metabolites is critical to understanding these complex interactions and developing microbiota-inspired therapies. Here, we use large-scale functional screening of molecules produced by individual members of a simplified human microbiota to identify bacterial metabolites that agonize G-protein-coupled receptors (GPCRs). Multiple metabolites, including phenylpropanoic acid, cadaverine, 9-10-methylenehexadecanoic acid, and 12-methyltetradecanoic acid, were found to interact with GPCRs associated with diverse functions within the nervous and immune systems, among others. Collectively, these metabolite-receptor pairs indicate that diverse aspects of human health are potentially modulated by structurally simple metabolites arising from primary bacterial metabolism.