Přejít k obsahu
Merck
Diversity-Oriented Synthesis of Steviol Glycosides.
The Journal of organic chemistry (2018-09-06)
Zhi Qiao, Hui Liu, Jing-Jing Sui, Jin-Xi Liao, Yuan-Hong Tu, Richard R Schmidt, Jian-Song Sun
ANOTACE
With cheap and easily available mixtures of steviol glycosides as starting materials, a practical method for steviol acquisition has been developed, on the basis of which a facile, diversity-oriented, and economic protocol for the synthesis of structurally defined steviol glycosides was established. The novel approach is featured by the highly efficient glycosylation of sterically hindered and acid-sensitive steviol via orchestrated application of Yu glycosylation, Schmidt glycosylation, and PTC glycosylation. Hence, these high-intensity sweeteners and potential lead compounds for drug development are now readily accessible.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Rebaudioside M, ≥95.0% (HPLC)