Přejít k obsahu
Merck
  • Exciplex Cohosts Employing Nonconjugated Linked Dicarbazole Donors for Highly Efficient Thermally Activated Delayed Fluorescence-Based Organic Light-Emitting Diodes.
Exciplex Cohosts Employing Nonconjugated Linked Dicarbazole Donors for Highly Efficient Thermally Activated Delayed Fluorescence-Based Organic Light-Emitting Diodes.
ACS applied materials & interfaces (2018-09-18)
Yi-Tzu Hung, Zhi-Yang Chen, Wen-Yi Hung, Deng-Gao Chen, Ken-Tsung Wong
ANOTACE
Two new nonconjugated linked dicarbazole materials, dCzPSi and dCzPSO2, with high triplet energy were synthesized and characterized. dCzPSi and dCzPSO2 were adopted as unipolar host materials for the green thermally activated delayed fluorescence (TADF) emitter (4CzIPN) to achieve high-efficiency organic light-emitting diodes (OLEDs). The electron-transporting acceptor, PO-T2T, was introduced to mix with dCzPSi and dCzPSO2 to give two new exciplex-forming systems that can improve the exciton formation propensity in the emitting layer. The relevant properties of these new exciplexes were characterized, and they were suggested as promising cohosts for the green TADF emitter, 4CzIPN. The characteristics of the devices employing single hosts (dCzPSi and dCzPSO2) and exciplex-forming cohosts (dCzPSi:PO-T2T and dCzPSO2:PO-T2T) were explored. The obtained results indicate that the Si-bridged dicarbazole compound dCzPSi outperforms its counterpart dCzPSO2 in which two carbazole groups are linked by an SO2 group. The device employed with the dCzPSi:PO-T2T cohost with 10 wt % 4CzIPN achieved a low Von of 2.2 V and maximum efficiencies of 21.1% (external quantum efficiency), 56.4 cd A-1 (current efficiency), 59.1 lm W-1 (power efficiency), as compared to those (18.7%, 56.6 cd A-1, and 68.5 lm W-1) of the dCzPSO2:PO-T2T-hosted device. This work verifies the advantages of using a cohost that can form an exciplex for boosting the device efficiency with reduced efficiency roll-off of TADF-based OLEDs.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
PO-T2T, >=99% (HPLC)