Přejít k obsahu
Merck
  • The involvement of ankyrin in the regulation of inositol 1,4,5-trisphosphate receptor-mediated internal Ca2+ release from Ca2+ storage vesicles in mouse T-lymphoma cells.
The involvement of ankyrin in the regulation of inositol 1,4,5-trisphosphate receptor-mediated internal Ca2+ release from Ca2+ storage vesicles in mouse T-lymphoma cells.
The Journal of biological chemistry (1993-04-05)
L Y Bourguignon, H Jin, N Iida, N R Brandt, S H Zhang
ANOTACE
Mouse T-lymphoma cells contain a unique type of internal vesicle which bands at the relatively light density of 1.07 g/cc. These vesicles do not contain any detectable Golgi, endoplasmic reticulum, plasma membrane, or lysosomal marker protein activities. Binding of [3H]inositol 1,4,5-trisphosphate (IP3) to these internal vesicles reveals the presence of a single, high affinity class of IP3 receptor with a dissociation constant (Kd) of 1.6 +/- 0.3 nM. Using a panel of monoclonal and polyclonal antibodies against IP3 receptor, we have established that the IP3 receptor (approximately 260 kDa) displays immunological cross-reactivity with the rat brain IP3 receptor. Polymerase chain reaction analysis of first-strand cDNAs from both mouse T-lymphoma cells and rat brain tissues reveals that the IP3 receptor transcript in mouse T-lymphoma cells belongs to the short form (non-neuronal form) and not the long form (neuronal form) detected in rat brain tissue. Scatchard plot analysis shows that high affinity binding occurs between ankyrin and the IP3 receptor with a Kd of 0.2 nM. Most importantly, the binding of ankyrin to the light density vesicles significantly inhibits IP3 binding and IP3-induced internal Ca2+ release. These findings suggest that the cytoskeleton plays a pivotal role in the regulation of IP3 receptor-mediated internal Ca2+ release during lymphocyte activation.