Přejít k obsahu
Merck
Mammalian Ras interacts directly with the serine/threonine kinase Raf.
Cell (1993-07-16)
A B Vojtek, S M Hollenberg, J A Cooper
ANOTACE
We have identified proteins that interact with H-Ras using a two hybrid system screen of a mouse cDNA library. Approximately 50% of the clones identified encoded portions of the c-Raf and A-Raf serine/threonine kinases. Overlaps among these clones define a conserved 81 residue region of the N-terminus of Raf as the Ras interaction region. We show that Raf interacts with wild-type and activated Ras, but not with an effector domain mutant of Ras or with a dominant-interfering Ras mutant. Using purified bacterially expressed fusion proteins, we show, furthermore, that Ras and the N-terminal region of Raf associate directly in vitro and that this interaction is dependent on GTP bound to Ras.