Přejít k obsahu
Merck
Regulation of protein synthesis by heme-regulated eIF-2 alpha kinase.
Trends in biochemical sciences (1995-03-01)
J J Chen, I M London
ANOTACE
Protein synthesis is regulated by the phosphorylation of the alpha-subunit of eukaryotic initiation factor 2 (eIF-2 alpha) in a variety of cells. At present, there are two distinct mammalian eIF-2 alpha kinases that have been cloned, the double-stranded-RNA-dependent eIF-2 alpha kinase (PKR) and the heme-regulated eIF-2 alpha kinase (HRI). HRI is activated under conditions of heme deficiency in immature erythroid cells, and its activity is inhibited by heme. The high levels of HRI in reticulocytes indicate that its major physiological role is the regulation of protein synthesis, particularly of hemoglobin, according to the concentration of heme in these cells.