Přejít k obsahu
Merck
Antibody-mediated rejection of cardiac allografts in CCR5-deficient recipients.
Journal of immunology (Baltimore, Md. : 1950) (2007-10-04)
Taiji Nozaki, Hiroyuki Amano, Alice Bickerstaff, Charles G Orosz, Andrew C Novick, Kazunari Tanabe, Robert L Fairchild
ANOTACE
Rejected MHC-mismatched cardiac allografts in CCR5(-/-) recipients have low T cell infiltration, but intense deposition of C3d in the large vessels and capillaries of the graft, characteristics of Ab-mediated rejection. The roles of donor-specific Ab and CD4 and CD8 T cell responses in the rejection of complete MHC-mismatched heart grafts by CCR5(-/-) recipients were directly investigated. Wild-type C57BL/6 and B6.CCR5(-/-) (H-2(b)) recipients of A/J (H-2(a)) cardiac allografts had equivalent numbers of donor-reactive CD4 T cells producing IFN-gamma, whereas CD4 T cells producing IL-4 were increased in CCR5(-/-) recipients. Numbers of donor-reactive CD8 T cells producing IFN-gamma were reduced 60% in CCR5(-/-) recipients. Day 8 posttransplant serum titers of donor-specific Ab were 15- to 25-fold higher in CCR5(-/-) allograft recipients, and transfer of this serum provoked cardiac allograft rejection in RAG-1(-/-) recipients within 14 days, whereas transfer of either serum from wild-type recipients or immune serum from CCR5-deficient recipients diluted to titers observed in wild-type recipients did not mediate this rejection. Wild-type C57BL/6 and B6.CCR5(-/-) recipients rejected A/J cardiac grafts by day 11, whereas rejection was delayed (day 12-60, mean 21 days) in muMT(-/-)/CCR5(-/-) recipients. These results indicate that the donor-specific Ab produced in CCR5(-/-) heart allograft recipients is sufficient to directly mediate graft rejection, and the absence of recipient CCR5 expression has differential effects on the priming of alloreactive CD4 and CD8 T cells.