Přejít k obsahu
Merck
Fas-induced caspase denitrosylation.
Science (New York, N.Y.) (1999-04-24)
J B Mannick, A Hausladen, L Liu, D T Hess, M Zeng, Q X Miao, L S Kane, A J Gow, J S Stamler
ANOTACE
Only a few intracellular S-nitrosylated proteins have been identified, and it is unknown if protein S-nitrosylation/denitrosylation is a component of signal transduction cascades. Caspase-3 zymogens were found to be S-nitrosylated on their catalytic-site cysteine in unstimulated human cell lines and denitrosylated upon activation of the Fas apoptotic pathway. Decreased caspase-3 S-nitrosylation was associated with an increase in intracellular caspase activity. Fas therefore activates caspase-3 not only by inducing the cleavage of the caspase zymogen to its active subunits, but also by stimulating the denitrosylation of its active-site thiol. Protein S-nitrosylation/denitrosylation can thus serve as a regulatory process in signal transduction pathways.