Přejít k obsahu
Merck
  • High-fat diet consumption reduces hepatic folate transporter expression via nuclear respiratory factor-1.
High-fat diet consumption reduces hepatic folate transporter expression via nuclear respiratory factor-1.
Journal of molecular medicine (Berlin, Germany) (2018-09-05)
Victoria Sid, Yaw L Siow, Yue Shang, Connie W Woo, Karmin O
ANOTACE
Folate is an essential micronutrient for biological function. The liver, a primary organ for folate metabolism and storage, plays an important role in folate homeostasis. Proton-coupled folate transporter (PCFT) and reduced folate carrier (RFC) are the major folate transporters responsible for folate uptake at basolateral membrane of hepatocytes. Low serum folate levels are frequently associated with obesity. We investigated the mechanism that regulated folate status in a mouse model with diet-induced obesity. Mice (C57BL/6J) were fed a high-fat diet (60% kcal fat) for 8 weeks. Mice displayed increased hepatic lipid accumulation and decreased folate levels in the liver and serum compared to mice fed a normal chow diet (10% kcal fat). High-fat diet-fed mice had low expression of PCFT and RFC and decreased nuclear respiratory factor-1 (NRF-1)/DNA-binding activity. Treatment with NRF-1 siRNA or palmitic acid reduced folate transporter expression in hepatocytes. Inhibition of NRF-1 mediated folate transporter expression significantly reduced intracellular folate levels. These results suggest that chronic consumption of high-fat diets impairs folate transporter expression via NRF-1-dependent mechanism, leading to reduced hepatic folate storage. Understanding the regulation of folate homeostasis in obesity may have an important implication in current guideline of folate intake. KEY MESSAGES: Serum and liver folate levels are decreased in diet-induced obese mice. Chronic high-fat diet consumption impairs expression of hepatic PCFT and RFC. NRF-1 regulates hepatic folate transporters expression and folate levels.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human NRF1