Přejít k obsahu
Merck
In Vitro Screening of Bioactive Compounds in some Gluten-Free Plants.
Applied biochemistry and biotechnology (2018-05-10)
Jerzy Drzewiecki, Alma Leticia Martinez-Ayala, María A Lozano-Grande, Hanna Leontowicz, Maria Leontowicz, Zenon Jastrzebski, Pawel Pasko, Shela Gorinstein
ANOTACE
Electrophoretic, antioxidant, and FTIR profiles of some varieties of amaranth, quinoa, and buckwheat seeds and their by products were compared. Water extracts of these products were evaluated by the Folin-Ciocalteau method in order to determine total phenolic content. The antioxidant activities were determined by 2,2'-azobis-2-methyl-propanimidamide, ferric-reducing/antioxidant power, and cupric reducing antioxidant capacity radical scavenging assays. FTIR spectra showed the secondary structure of pseudocereals in the ranges of amides I, II, and III shifts. Results of evaluated methods could be used to control several products (seeds, flours, extracts, flakes, roasting) with high phenolic content and antioxidant activity suitable for supplementation in food applications. Graphical Abstract ᅟ.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
s-Triazine, 97%