Přejít k obsahu
Merck
An odorant receptor from Anopheles sinensis in China is sensitive to oviposition attractants.
Malaria journal (2018-10-07)
Hongmei Liu, Luhong Liu, Peng Cheng, Xiaodan Huang, Maoqing Gong
ANOTACE
Anopheles sinensis is an important vector for the spread of malaria in China. Olfactory-related behaviours, particularly oviposition site seeking, offer opportunities for disrupting the disease-transmission process. This is the first report of the identification and characterization of AsinOrco and AsinOR10 in An. sinensis. AsinOrco and AsinOR10 share 97.49% and 90.37% amino acid sequence identity, respectively, with related sequences in Anopheles gambiae. A functional analysis demonstrated that AsinOrco- and AsinOR10-coexpressing HEK293 cells were highly sensitive to 3-methylindole, but showed no significant differences in response to other test odorants when compared to DMSO. AsinOrco was characterized as a new member of the Orco ortholog subfamily. AsinOR10, which appears to be a member of the OR2-10 subfamily, is directly involved in identification of oviposition sites. This finding will help to elucidate the molecular mechanisms underlying olfactory signaling in An. sinensis and provide many more molecular targets for eco-friendly pest control.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
1-Methylindole, ≥97%