Přejít k obsahu
Merck
Reprogramming of the antimycin NRPS-PKS assembly lines inspired by gene evolution.
Nature communications (2018-09-01)
Takayoshi Awakawa, Takuma Fujioka, Lihan Zhang, Shotaro Hoshino, Zhijuan Hu, Junko Hashimoto, Ikuko Kozone, Haruo Ikeda, Kazuo Shin-Ya, Wen Liu, Ikuro Abe
ANOTACE
Reprogramming of the NRPS/PKS assembly line is an attractive method for the production of new bioactive molecules. However, it is usually hampered by the loss of intimate domain/module interactions required for the precise control of chain transfer and elongation reactions. In this study, we first establish heterologous expression systems of the unique antimycin-type cyclic depsipeptides: JBIR-06 (tri-lactone) and neoantimycin (tetra-lactone), and engineer their biosyntheses by taking advantage of bioinformatic analyses and evolutionary insights. As a result, we successfully accomplish three manipulations: (i) ring contraction of neoantimycin (from tetra-lactone to tri-lactone), (ii) ring expansion of JBIR-06 (from tri-lactone to tetra-lactone), and (iii) alkyl chain diversification of JBIR-06 by the incorporation of various alkylmalonyl-CoA extender units, to generate a set of unnatural derivatives in practical yields. This study presents a useful strategy for engineering NRPS-PKS module enzymes, based on nature's diversification of the domain and module organizations.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
(S)-(−)-2-Hydroxyisocaproic acid, 98%