Přejít k obsahu
Merck
  • c-Jun dimerizes with itself and with c-Fos, forming complexes of different DNA binding affinities.
c-Jun dimerizes with itself and with c-Fos, forming complexes of different DNA binding affinities.
Cell (1988-12-02)
T D Halazonetis, K Georgopoulos, M E Greenberg, P Leder
ANOTACE
The c-Jun and c-fos proto-oncogenes encode proteins that form a complex which regulates transcription from promoters containing AP-1 activation elements. c-Jun has specific DNA binding activity, while c-Fos has homology to the putative DNA binding domain of c-Jun. Following in vitro translation, c-Jun binds as a homodimer to the AP-1 DNA site, while c-Fos fails to dimerize and displays no apparent affinity for the AP-1 element. Cotranslated c-Jun and c-Fos proteins bind 25 times more efficiently to the AP-1 DNA site as a heterodimer than does the c-Jun homodimer. These experiments suggest that in growth factor-stimulated cells c-Jun binds DNA as a dimer with c-Fos as its natural partner. However, overexpression of c-Jun protein in the absence of c-Fos may result in formation of aberrant homodimeric transcription complexes, which could abrogate the normal mechanisms controlling gene expression.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
c-JUN (1-79), GST tagged human, recombinant, expressed in E. coli, ≥50% (SDS-PAGE), buffered aqueous glycerol solution