Přejít k obsahu
Merck
The TNF family members BAFF and APRIL: the growing complexity.
Cytokine & growth factor reviews (2003-06-06)
Fabienne Mackay, Christine Ambrose
ANOTACE
B cell activating factor belonging to the TNF family (BAFF) and apoptosis-inducing ligand (APRIL) are two related members of the TNF ligand superfamily. Although they share two receptors, TACI and BCMA, transgenic and knockout mice in this system reveal that their functions are not redundant. BAFF is a critical survival/maturation factor for peripheral B cells and this activity is mediated through a BAFF-specific receptor, BAFF-R. Overexpression of BAFF has been linked to autoimmune disease and aspects of B cell neoplasia. APRIL appears to play a role in T-independent type II antigen responses and T cell survival, but can also induce proliferation/survival of non-lymphoid cells. Elevated expression of APRIL has been found in some tumor cell lines and in tumor tissue libraries. Therapies designed to inhibit the BAFF and APRIL pathways holds great promise for the future.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
APRIL human, recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
APRIL from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture