Přejít k obsahu
Merck
E2F1 inhibition mediates cell death of metastatic melanoma.
Cell death & disease (2018-05-11)
Florian Rouaud, Nedra Hamouda-Tekaya, Michaël Cerezo, Patricia Abbe, Joséphine Zangari, Veronique Hofman, Mickaël Ohanna, Baharia Mograbi, Najla El-Hachem, Zohra Benfodda, Alexandre Lebeau, Meri K Tulic, Paul Hofman, Corine Bertolotto, Thierry Passeron, Jean-Sébastien Annicotte, Robert Ballotti, Stéphane Rocchi
ANOTACE
Melanoma is one of the most lethal cancers when it reaches a metastatic stage. Despite advancements in targeted therapies (BRAF inhibitors) or immunotherapies (anti-CTLA-4 or anti-PD1), most patients with melanoma will need additional treatment. Thus, there is an urgent need to develop new therapeutical approaches to bypass resistance and achieve more prolonged responses. In this context, we were interested in E2F1, a transcription factor that plays a major role in the control of cell cycle under physiological and pathological conditions. Here we confirmed that E2F1 is highly expressed in melanoma cells. Inhibition of E2F1 activity further increased melanoma cell death and senescence, both in vitro and in vivo. Moreover, blocking E2F1 also induced death of melanoma cells resistant to BRAF inhibitors. In conclusion, our studies suggest that targeting the E2F1 signaling pathway may be therapeutically relevant for melanoma.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
HLM006474, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human E2F1