Přejít k obsahu
Merck
  • Evaluation of alternatives to cautery disbudding of dairy goat kids using physiological measures of immediate and longer-term pain.
Evaluation of alternatives to cautery disbudding of dairy goat kids using physiological measures of immediate and longer-term pain.
Journal of dairy science (2018-03-27)
Melissa N Hempstead, Joseph R Waas, Mairi Stewart, Vanessa M Cave, Mhairi A Sutherland
ANOTACE
We evaluated alternatives to cautery disbudding of goat kids using physiological measures of immediate and longer-term pain. Fifty Saanen doe kids were randomly assigned to 1 of 5 treatments (n = 10/treatment): (1) cautery disbudding (CAUT), (2) caustic paste disbudding (CASP), (3) liquid nitrogen disbudding (CRYO), (4) clove oil injected into the horn bud (CLOV), or (5) sham disbudding (SHAM). Serum cortisol and haptoglobin concentrations were measured from blood samples collected immediately before treatment (baseline) and at 15, 30, 60, and 120 min and then again at 6 and 24 h post-treatment. An infrared thermography camera was used to take images of the horn buds 24 h pre- and 24, 48, and 72 h post-treatment to measure skin temperature. Body weight was measured daily for 1 wk to assess weight change post-treatment. Images of the horn buds were taken at d 1, 2, and 7 and at 6 wk post-treatment to assess tissue damage and wound healing. Mean cortisol concentrations were elevated in CASP kids 1 h post-treatment relative to CAUT kids. Cortisol concentrations of CRYO kids were higher than those of CAUT kids 30 min post-treatment; concentrations for CLOV kids were similar to CAUT kids post-treatment. Mean haptoglobin concentrations were similar across treatments over time; however, CLOV kids had higher concentrations at 24 h post-treatment than all other treatments. Skin temperatures of CASP and CLOV kids were elevated relative to CAUT kids at all time points post-treatment, and all disbudded kids had skin temperatures above those of SHAM kids at 72 h post-treatment. Treatment did not influence weight gain. The CAUT kids had large, open wounds exposing bone; small scabs were still evident 6 wk post-treatment. The CASP kids had red and open, raw wounds that generated large eschars, apparent for up to 6 wk. The CRYO kids had closed, dry wounds initially, but over time lesions appeared that caused open wounds; small scabs were present 6 wk post-treatment. The CLOV kids had closed, dry wounds with blackened skin; healed skin and minimal scabs were present 6 wk post-treatment. Caustic paste and cryosurgical disbudding appeared to cause more pain compared with cautery disbudding; thus, these methods may not provide good alternatives to cautery disbudding. Clove oil appeared to cause a similar pain response as cautery disbudding and smaller wounds with earlier tissue repair; this method shows promise as an alternative to cautery disbudding.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Penta-O-galloyl-β-D-glucose hydrate, ≥96% (HPLC)
Sigma-Aldrich
Clove oil