Zpět
Water sensor
Products
Kategorie produktu
sensors (6)
Značka
Milli-Q (6)
Průměr (mm)
Délka (m)
Kompatibilita
Filtrovat produkty
Showing 1-6 of 6
Porovnat
Číslo produktu
Popis
Stanovení ceny
ZFWATDET2
Water sensor,
Detects water spillage on the floor and automatically closes the stand-alone inlet solenoid valve., AC/DC input 230 V, 50–60 Hz
Rozšířit
Zobrazit stanovené ceny
ZWATSENA1
Water sensor,
Safety accessory that detects a water leak, for use with Milli-Q
®
IQ/IX/EQ/SQ series.
Rozšířit
Zobrazit stanovené ceny
TANKLK001
Water sensor,
For use with RiOs
™
and Elix
®
water purification systems
Rozšířit
Zobrazit stanovené ceny
ZFWATDET4
Water sensor,
For use with AFS, Elix Advantage / Essential / Reference, Milli-Q Direct / Reference and RiOs Essential systems
Rozšířit
Zobrazit stanovené ceny
ZFWATDET1
Water sensor,
Detects water spillage on the floor and automatically closes the stand-alone inlet solenoid valve., AC/DC input 120 V, 60 Hz
Rozšířit
Zobrazit stanovené ceny
TANKLKXL1
Water sensor,
For high volume water purification systems