Zpět
Vent Filter
Products
Kategorie produktu
filters (12)
Značka
Milli-Q (12)
Průměr (mm)
Délka (palce)
Délka (mm)
Kompatibilita
Úroveň kvality
Hydrofobicita
Materiál pouzdra
Velikost pórů (µm)
Filtrovat produkty
Showing 1-12 of 12
Porovnat
Číslo produktu
Popis
Stanovení ceny
TANKMPK01
Vent Filter,
Protects purified water stored in 30/60/100 PE tanks from airborne contaminants.
Rozšířit
Zobrazit stanovené ceny
TANKV01A1
Vent Filter,
Protects pure water in Milli-Q
®
IQ/IX/EQ storage tanks from airborne contaminants
Rozšířit
Zobrazit stanovené ceny
ZFRE012FC
Vent Filter,
Protects stored pure water quality by providing advanced protection against airborne contaminants.
Rozšířit
Zobrazit stanovené ceny
TANKMPK02
Vent Filter,
Protects purified water stored in 30/60/100 PE tanks from airborne contaminants.
Rozšířit
Zobrazit stanovené ceny
TANKMPK03
Vent Filter,
Prevents contamination of pure water stored in 6/10 L internal storage tanks
Rozšířit
Zobrazit stanovené ceny
TANKVNT02
Vent Filter,
Prevents contamination of pure water in SDS storage systems
Rozšířit
Zobrazit stanovené ceny
TANKMPKXC
Vent Filter,
Advanced protection against airborne contaminants for storage tanks of Milli-Q
®
CLX 7000 series systems
Rozšířit
Zobrazit stanovené ceny
SLFH02510
Vent Filter,
For use with Direct-Q
®
(1997-2010) / AFS
®
(2003-2012) / Simplicity
®
systems with integrated storage tanks of 2 / 3.5 / 10 L capacity
Rozšířit
Zobrazit stanovené ceny
TANKVH1A1
Vent Filter,
Protects pure water in Milli-Q
®
IQ/IX/EQ storage tanks from airborne contaminants. For High-Flow applications.
Rozšířit
Zobrazit stanovené ceny
ZFRE012F0
Vent Filter,
Prevents contamination of pure water stored in SDS tanks
Rozšířit
Zobrazit stanovené ceny
TANKVNT22
Vent Filter,
pore size 0.22 μm, carbon, and soda lime, Advanced protection against airborne contaminants for storage tanks
Rozšířit
Zobrazit stanovené ceny
TANKVNT01
Vent Filter,
Prevents contamination of pure water in SDS storage systems