Merck
Biopak® Polisher Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q® IQ/IX/EQ systems
Biopak® Polisher
Biopak® Polisher Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q® IQ/IX/EQ systems
Products
Kategorie produktu
Značka
Kompatibilita