Merck
3D Biotek Polystyrene (PS) scaffold inserts for 3D bioreactor 24 well chamber, fiber diam. ~150 μm, ~ 200 μm spacing
3D Biotek Polystyrene (PS) scaffold inserts
3D Biotek Polystyrene (PS) scaffold inserts for 3D bioreactor 24 well chamber, fiber diam. ~150 μm, ~ 200 μm spacing
Products
Kategorie produktu
Výrobce
Typ vazby
Velikost pórů (µm)
Sterilita
Showing 1-1 of 1