57041
Vlastnosti
100
pk of 12
solid phase extraction (SPE): suitable
food and beverages
Popis
Zobrazit rozdíly
1 of 1
Supelco
Retaining Clips For Visiprep™ SPE Manifold Collection Racks
57037
Visiprep™ Manifold Collection Rack (included with standard and DL models)
57255
57022
Empty SPE Tube (no frits)
included with standard and DL, 12- and 24-port models, pk of 12
for use with 12-port model, includes base, 3 support rods, center plate, plate for 10 mm test tubes, 12 retaining clips
for use with 24-port model, includes base, 2 support rods, center plate, plate for 10 mm test tubes, 8 retaining clips
volume 60 mL, pk of 12
PFAS testingfood and beverages
-
Vyberte jednu z posledních verzí:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.