Přeskočit na
22949
Union, reducing
zero dead volume, for connecting 1/8 to 1/16 in., bore size 0.25 mm
Synonyma:
Valco® Unions
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
Vám/Skladová položkaDostupnostCena
1 ea
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
689,00 Kč
O této položce
UNSPSC Code:
41115700
NACRES:
NB.21
689,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Požádejte o zakázkovou objednávku
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® IZR21C
bore size
0.25 mm
fitting
1/16 in. Valco (nut and ferrule included)
compatibility
for connecting 1/8 to 1/16 in.
General description
Stainless steel, unless noted otherwise. Nuts and ferrules included.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Porovnat podobné položky
Zobrazit úplné porovnání
Zobrazit rozdíly
1 of 1
Tato položka
59025-U5824922999
Union, reducing zero dead volume, for connecting 1/8 to 1/16 in., bore size 0.25 mm
Supelco
22949
Union, reducing
Union, reducing for connecting 1/16 to 1/32 in., bore size 0.25 mm
59025-U
Union, reducing
vibrant-m
58249
Valco® Unions
Valco® Unions internal reducing, bore 0.75 mm, for connecting 1/8 to 1/16 in.
Supelco
22999
Valco® Unions
description
zero dead volume, for connecting 1/8 to 1/16 in., bore size 0.25 mm
description
for connecting 1/16 to 1/32 in., bore size 0.25 mm
description
internal reducing, 1/4 to 1/16 in., bore size 0.75 mm
description
internal reducing, bore 0.75 mm, for connecting 1/8 to 1/16 in.
fitting
1/16 in. Valco (nut and ferrule included)
fitting
10-32 male Valco for 1/16 in. O.D. tubing, 6-32 Valco for 1/32 in. O.D. tubing
fitting
1/16 in. Valco (nut and ferrule included), 1/4 in. Valco (nut and ferrule included)
fitting
1/16 in. Valco, 1/8 in. Valco
bore size
0.25 mm
bore size
0.25 mm
bore size
0.75 mm
bore size
-
material
stainless steel
material
stainless steel union and fittings (nuts and ferrules included)
material
stainless steel
material
stainless steel (nuts and ferrules included)
manufacturer/tradename
VICI Valco® IZR21C
manufacturer/tradename
-
manufacturer/tradename
-
manufacturer/tradename
-
compatibility
for connecting 1/8 to 1/16 in.
compatibility
for connecting 1/16 to 1/32 in.
compatibility
1/4 to 1/16 in.
compatibility
for connecting 1/8 to 1/16 in.
packaging
pkg of 1 ea
packaging
pkg of 1 unit
packaging
pkg of 1 ea
packaging
pkg of 1 ea
Still not finding the right product?
Explore all of our products under Union, reducing
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)