Yes, this product is compatible with Shimadzu 8010-0263.
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
50 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 6 690,00 Kč |
O této položce
UNSPSC Code:
41115717
NACRES:
SB.54
Packaging:
pkg of 50 ea
6 690,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlisize
(plug type)
packaging
pkg of 50 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
diam. × L
~6 mm × 9 mm
compatibility
for use with Shimadzu all
General description
Septum is a very important component of syringe injection system. They are usually fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene or fluoroelastomers. It is suitable to conveniently introducing the sample without causing the system to leak. Improper septa decomposes under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
For use in Shimadzu GCs that require plug septa.
Application
Thermogreen® LB-2 Septa is suitable for the following applications:
- To minimize extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS.
- Thermo-resistant septa is used in GC instrument to avoid carrier gas leaks, extraneous peaks and phthalate contamination.[1]
- Thermo-resistant septa along with thick needle protector is used in GC instruments to avoid damage irreversibly caused by microsyringe.[2]
- It is also used in the influent sampling ports in gas chromatographs.
Other Notes
For a full listing and description of our GC Inlet Septa (click here)
Legal Information
Thermogreen is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
Tato položka | |||
|---|---|---|---|
| description Shimadzu plug, pkg of 50 ea | description diam. 11 mm ( 7/16 in.) | description Shimadzu plug, pkg of 10 ea | description diam. 12.5 mm ( 1/2 in.) |
| compatibility for use with Shimadzu all | compatibility - | compatibility for use with Shimadzu all | compatibility - |
| packaging pkg of 50 ea | packaging pkg of 50 pkg | packaging pkg of 10 ea | packaging pkg of 50 ea |
| size (plug type) | size - | size (plug type) | size - |
| diam. × L ~6 mm × 9 mm | diam. × L - | diam. × L ~6 mm × 9 mm | diam. × L - |
Still not finding the right product?
Explore all of our products under Thermogreen® LB-2 Septa, plug
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
-
Is this product compatible with shimadzu 8010-0263?
1 answer-
Helpful?
-
-
Is these septa fit the outlet 1/8''?
1 answer-
This septa has a diameter of approximately 6 mm and would not be suitable for a 1/8 in. (3.175 mm) outlet.