Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|
O této položce
UNSPSC Code:
12352200
NACRES:
SB.54
Packaging:
pkg of 10 ea
Informace o stanovených cenách a dostupnosti nejsou v současnosti dostupné.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlisize
(plug type)
packaging
pkg of 10 ea
technique(s)
gas chromatography (GC): suitable
diam. × L
~6 mm × 9 mm
compatibility
for use with Shimadzu all
General description
Septum is a very important component of syringe injection system. They are usually fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene or fluoroelastomers. It is suitable to conveniently introducing the sample without causing the system to leak. Improper septa decomposes under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
For use in Shimadzu GCs that require plug septa.
Application
Thermogreen® LB-2 Septa is suitable for the following applications:
- To minimize extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS.
- Thermo-resistant septa is used in GC instrument to avoid carrier gas leaks, extraneous peaks and phthalate contamination.[1]
- Thermo-resistant septa along with thick needle protector is used in GC instruments to avoid damage irreversibly caused by microsyringe.[2]
- It is also used in the influent sampling ports in gas chromatographs.
Other Notes
For a full listing and description of our GC Inlet Septa (click here)
Legal Information
Thermogreen is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
Tato položka | |||
|---|---|---|---|
| compatibility for use with Shimadzu all | compatibility - | compatibility for use with Shimadzu all | compatibility - |
| packaging pkg of 10 ea | packaging pkg of 50 pkg | packaging pkg of 50 ea | packaging pkg of 50 ea |
| technique(s) gas chromatography (GC): suitable | technique(s) - | technique(s) - | technique(s) - |
| size (plug type) | size - | size (plug type) | size - |
| diam. × L ~6 mm × 9 mm | diam. × L - | diam. × L ~6 mm × 9 mm | diam. × L - |
Still not finding the right product?
Explore all of our products under Thermogreen® LB-2 Septa, plug
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?