O této položce
NACRES:
SE.11
UNSPSC Code:
41115418
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimanufacturer/tradename
Benchmark Scientific MR9600-590
General description
Absorbancy filters for the Accuris SmartReader plate reader.
Application
Microplate absorbance reader filters are used in a wide range of applications;
- ELISA (Enzyme-Linked Immunosorbent Assays)
- Protein Quantification techniques such as the Bradford assay
- DNA / RNA quantification
- Cell viability and proliferation assays
- Enzyme kinetics
- High-throughput screening
- Drug target validation
- Infectious disease detection
- Food and Beverage contamination detection
- Environmental monitoring
- Qualification of expression of engineered proteins in synthetic biology
- Quality control in many manufacturing processes.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
SmartReader is a trademark of Benchmark Scientific, Inc.
Still not finding the right product?
Explore all of our products under Accuris™ SmartReader™ 96 microplate absorbancy reader filter
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| Z742736-1EA | 04061835331406 |