Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 51 000,00 Kč |
O této položce
51 000,00 Kč
material
stainless steel
feature
stainless steel rotor/stator
packaging
pack of 1
manufacturer/tradename
IKA 0001024200
diam.
8 mm
General description
Features and Benefits
- suitable for use with solvents
- sterilization methods: all methods
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description dispersing element for T-25 digital disperser, 8 mm, 1/cs | description dispersing for T-25 digital disperser, 10 mm, 1/cs | description dispersing element for T-25 digital disperser, 18 mm, 1/cs | description dispersing element for T-10 basic disperser, 10 mm, 1/cs |
| manufacturer/tradename IKA 0001024200 | manufacturer/tradename IKA 0000594000 | manufacturer/tradename IKA 0000593400 | manufacturer/tradename IKA 3370100 |
| material stainless steel | material stainless steel | material stainless steel | material stainless steel |
| packaging pack of 1 | packaging pack of 1 | packaging pack of 1 | packaging pack of 1, case of 1 |
| feature stainless steel rotor/stator | feature stainless steel rotor/stator | feature stainless steel rotor/stator | feature stainless steel rotor/stator |
| diam. 8 mm | diam. 10 mm | diam. 18 mm | diam. 10 mm |
Still not finding the right product?
Explore all of our products under IKA® ULTRA-TURRAX® disperser tools, stainless steel
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?